1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480145-1G
APOSOR480145-1GEA 4540 EUR
APOSOR480145-1G
1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496586
CAS-Nummer: 1780678-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR