1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480149-1G
APOSOR480149-1GEA 4540 EUR
APOSOR480149-1G
1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid
1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496592
CAS-Nummer: 1785441-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR