1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480151-1G
APOSOR480151-1GEA 4540 EUR
APOSOR480151-1G
1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid
1-(8-Bromonaphthalen-2-yl)cyclopropane-1-carboxylic acid
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496594
CAS-Nummer: 1781365-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR