1-(7-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Lieferant: Apollo Scientific
OR480153-1G
APOSOR480153-1GEA
4540
EUR
APOSOR480153-1G
1-(7-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
1-(7-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Formel:
C₁₄H₁₀BrN Molecular Weight: 272,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496596 CAS-Nummer: 2121970-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...