1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Lieferant: Apollo Scientific
OR480154-1G
APOSOR480154-1GEA
4540
EUR
APOSOR480154-1G
1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
1-(5-Bromonaphthalen-2-yl)cyclopropane-1-carbonitrile
Formel:
C₁₄H₁₀BrN Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496597 CAS-Nummer: 2116763-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...