1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile

Lieferant: Apollo Scientific

OR480161-1G
APOSOR480161-1GEA 4540 EUR
APOSOR480161-1G
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
Formel: C₁₄H₁₀BrN
Molecular Weight: 272,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496523
CAS-Nummer: 2113847-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR