1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile

Lieferant: Apollo Scientific

OR480163-1G
APOSOR480163-1GEA 4540 EUR
APOSOR480163-1G
1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
Formel: C₁₄H₁₀BrN
Molecular Weight: 272,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496529
CAS-Nummer: 1800258-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR