1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile

Lieferant: Apollo Scientific

OR480164-1G
APOSOR480164-1GEA 4540 EUR
APOSOR480164-1G
1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
1-(8-Bromonaphthalen-1-yl)cyclopropane-1-carbonitrile
Formel: C₁₄H₁₀BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD30496532
CAS-Nummer: 2114654-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR