1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine
Lieferant: Apollo Scientific
OR480165-1G
APOSOR480165-1GEA
4540
EUR
APOSOR480165-1G
1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine
1-(8-Bromonaphthalen-1-yl)cyclopropan-1-amine
Formel:
C₁₃H₁₂BrN Molecular Weight: 262,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496535 CAS-Nummer: 1780001-29-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...