1-(3-Bromonaphthalen-2-yl)cyclopropan-1-amine

Lieferant: Apollo Scientific

OR480173-1G
APOSOR480173-1GEA 4540 EUR
APOSOR480173-1G
1-(3-Bromonaphthalen-2-yl)cyclopropan-1-amine
1-(3-Bromonaphthalen-2-yl)cyclopropan-1-amine
Formel: C₁₃H₁₂BrN
Molecular Weight: 262,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496555
CAS-Nummer: 1704104-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR