1-(6-Bromonaphthalen-2-yl)cyclopropan-1-amine

Lieferant: Apollo Scientific

OR480176-1G
APOSOR480176-1GEA 4540 EUR
APOSOR480176-1G
1-(6-Bromonaphthalen-2-yl)cyclopropan-1-amine
1-(6-Bromonaphthalen-2-yl)cyclopropan-1-amine
Formel: C₁₃H₁₂BrN
Molecular Weight: 262,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496564
CAS-Nummer: 1704128-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR