1-(7-Bromonaphthalen-2-yl)cyclopropan-1-amine
Lieferant: Apollo Scientific
OR480177-1G
APOSOR480177-1GEA
4540
EUR
APOSOR480177-1G
1-(7-Bromonaphthalen-2-yl)cyclopropan-1-amine
1-(7-Bromonaphthalen-2-yl)cyclopropan-1-amine
Formel:
C₁₃H₁₂BrN Molecular Weight: 262,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496567 CAS-Nummer: 1782314-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...