DL-threo-3-Phenylserine hydrate 98%

Lieferant: Apollo Scientific

OR480205-5G
APOSOR480205-5GEA 105 EUR
APOSOR480205-5G
DL-threo-3-Phenylserine hydrate 98%
DL-threo-3-Phenylserine hydrate
Formel: C₉H₁₁NO₃·H₂O
Molecular Weight: 199,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863724
CAS-Nummer: 207605-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR