2-Tetrahydrofuran-2-ylacetic acid

Lieferant: Apollo Scientific

OR480227-1G OR480227-250MG OR480227-500MG
APOSOR480227-1GEA 570 EUR
APOSOR480227-1G APOSOR480227-250MG APOSOR480227-500MG
2-Tetrahydrofuran-2-ylacetic acid
2-Tetrahydrofuran-2-ylacetic acid
Formel: C₆H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03990566
CAS-Nummer: 2434-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR