Methyl-4-amino-1-boc-piperidine-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-Boc-4-Aminopiperidine-4-carboxylate

OR480232-1G OR480232-500MG OR480232-5G
APOSOR480232-1GEA 99.2 EUR
APOSOR480232-1G APOSOR480232-500MG APOSOR480232-5G
Methyl-4-amino-1-boc-piperidine-4-carboxylate 95%
Methyl-4-amino-1-boc-piperidine-4-carboxylate
Formel: C₁₂H₂₂N₂O₄
Molecular Weight: 258.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06200674
CAS-Nummer: 321997-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR