2-Chlor-3-formyl-6-picolin 97%

Lieferant: Apollo Scientific

OR480236-5G
APOSOR480236-5GEA 136 EUR
APOSOR480236-5G
2-Chlor-3-formyl-6-picolin 97%
2-Chlor-3-formyl-6-picolin
Formel: C₇H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD07368878
CAS-Nummer: 91591-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR