Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Methoxycarbonyl)cyclopropanecarboxylic acid

OR480237-100G
APOSOR480237-100GEA 102 EUR
APOSOR480237-100G
Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Cyclopropane-1,1-dicarboxylic acid mono methyl ester
Formel: C₆H₈O₄
Molecular Weight: 144.13 g/mol
Siedepunkt: 85…87 °C
Schmelzpunkt: 47…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06800496
CAS-Nummer: 113020-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR