1-(2-Nitrophenyl)-1H-pyrazole 97%

Lieferant: Apollo Scientific

OR480243-10G OR480243-1G OR480243-5G
APOSOR480243-10GEA 674 EUR
APOSOR480243-10G APOSOR480243-1G APOSOR480243-5G
1-(2-Nitrophenyl)-1H-pyrazole 97%
1-(2-Nitrophenyl)-1H-pyrazole
Formel: C₉H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06661759
CAS-Nummer: 25688-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR