2-Bromo-5-isopropylthiazole 95%

Lieferant: Apollo Scientific

OR480272-1G OR480272-250MG OR480272-500MG
APOSOR480272-1GEA 91.9 EUR
APOSOR480272-1G APOSOR480272-250MG APOSOR480272-500MG
2-Bromo-5-isopropylthiazole 95%
2-Bromo-5-isopropylthiazole
Formel: C₆H₈BrNS
Molecular Weight: 206.103 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12033480
CAS-Nummer: 1159817-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR