2-Bromo-5-isopropylthiazole 95%
Lieferant: Apollo Scientific
OR480272-1G
OR480272-250MG
OR480272-500MG
APOSOR480272-1GEA
91.9
EUR
APOSOR480272-1G
APOSOR480272-250MG
APOSOR480272-500MG
2-Bromo-5-isopropylthiazole 95%
2-Bromo-5-isopropylthiazole
Formel:
C₆H₈BrNS Molecular Weight: 206.103 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12033480 CAS-Nummer: 1159817-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...