(E)-4-(3-Butoxyacryloyl)benzonitrile
Lieferant: Apollo Scientific
OR480287-1G
OR480287-250MG
OR480287-500MG
APOSOR480287-1GEA
861
EUR
APOSOR480287-1G
APOSOR480287-250MG
APOSOR480287-500MG
(E)-4-(3-Butoxyacryloyl)benzonitrile
(E)-4-(3-Butoxyacryloyl)benzonitrile
Formel:
C₁₄H₁₅NO₂ Molecular Weight: 229.274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22683247 CAS-Nummer: 1379801-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...