1-Methyl-DL-tryptophan 95%

Lieferant: Apollo Scientific

OR480297-1G
APOSOR480297-1GEA 0 EUR
APOSOR480297-1G
1-Methyl-DL-tryptophan 95%
1-Methyl-DL-tryptophan
Formel: C₁₂H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00005803
CAS-Nummer: 26988-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR