2-Amino-3-(3-bromophenyl)propanoic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-DL-phenylalanine, H-DL-Phe(3-Br)-OH

OR480301-5G
APOSOR480301-5GEA 85.7 EUR
APOSOR480301-5G
2-Amino-3-(3-bromophenyl)propanoic acid 98%
2-Amino-3-(3-bromophenyl)propanoic acid
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05227935
CAS-Nummer: 30163-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR