4-Brom-DL-phenylalanin 97%
Lieferant: Apollo Scientific
OR480302-10G
OR480302-25G
APOSOR480302-10GEA
116
EUR
APOSOR480302-10G
APOSOR480302-25G
4-Brom-DL-phenylalanin 97%
4-Brom-DL-phenylalanin
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002599 CAS-Nummer: 14091-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...