4-Brom-DL-phenylalanin 97%

Lieferant: Apollo Scientific

OR480302-25G
APOSOR480302-25GEA 0 EUR
APOSOR480302-25G
4-Brom-DL-phenylalanin 97%
4-Brom-DL-phenylalanin
Formel: C₉H₁₀BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002599
CAS-Nummer: 14091-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR