3-(3,4-Dihydroxyphenyl)-DL-alanin 96%

Lieferant: Apollo Scientific

Synonyme: DL-DOPA

OR480320-100G
APOSOR480320-100GEA 360 EUR
APOSOR480320-100G
3-(3,4-Dihydroxyphenyl)-DL-alanin 96%
3-(3,4-Dihydroxyphenyl)-DL-alanin
Formel: C₉H₁₁NO₄
Molecular Weight: 197,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063060
CAS-Nummer: 63-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR