4-Amino-1-benzylpiperidine-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR480328-1G
APOSOR480328-1GEA 59.6 EUR
APOSOR480328-1G
4-Amino-1-benzylpiperidine-4-carboxylic acid 95%
4-Amino-1-benzylpiperidine-4-carboxylic acid
Formel: C₁₃H₁₈N₂O₂
Molecular Weight: 234,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794952
CAS-Nummer: 39143-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR