Ethyl-2-bromo-6-benzothiazolecarboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-bromo-6-benzothiazolecarboxylate

OR480384-1G
APOSOR480384-1GEA 71.2 EUR
APOSOR480384-1G
Ethyl-2-bromo-6-benzothiazolecarboxylate 97%
Ethyl-2-bromo-6-benzothiazolecarboxylate
Formel: C₁₀H₈BrNO₂S
Molecular Weight: 286,1500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669850
CAS-Nummer: 99073-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR