(2S)-2-Amino-2-(3-chlorophenyl)acetic acid 97%

Lieferant: Apollo Scientific

OR480389-1G
APOSOR480389-1GEA 109 EUR
APOSOR480389-1G
(2S)-2-Amino-2-(3-chlorophenyl)acetic acid 97%
(2S)-2-Amino-2-(3-chlorophenyl)acetic acid
Formel: C₈H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03839814
CAS-Nummer: 119565-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR