(S)-2-Amino-3-(3-methoxyphenyl)propanoic acid 97%

Lieferant: Apollo Scientific

OR480391-1G
APOSOR480391-1GEA 92.8 EUR
APOSOR480391-1G
(S)-2-Amino-3-(3-methoxyphenyl)propanoic acid 97%
(S)-2-Amino-3-(3-methoxyphenyl)propanoic acid
Formel: C₁₀H₁₃NO₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 33879-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR