2-Amino-2-cyclohexylacetic acid 97%

Lieferant: Apollo Scientific

OR480392-25G
APOSOR480392-25GEA 58 EUR
APOSOR480392-25G
2-Amino-2-cyclohexylacetic acid 97%
2-Amino-2-cyclohexylacetic acid
Formel: C₈H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05662746
CAS-Nummer: 5664-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR