3-Phenyl-L-serine 95%

Lieferant: Apollo Scientific

Synonyme: L-threo-Phenylserine

OR480411-1G
APOSOR480411-1GEA 400 EUR
APOSOR480411-1G
3-Phenyl-L-serine 95%
3-Phenyl-L-serine
Formel: C₉H₁₁NO₃
Molecular Weight: 181,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671668
CAS-Nummer: 6254-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR