4,4'-Disulphanediylbis(2-aminobutanoic acid) 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-[(3-amino-4-hydroxy-4-oxo-butyl)disulphanyl]butanoic acid

OR480422-100G
APOSOR480422-100GEA 208 EUR
APOSOR480422-100G
4,4'-Disulphanediylbis(2-aminobutanoic acid) 95+%
4,4'-Disulphanediylbis(2-aminobutanoic acid)
Formel: C₈H₁₆N₂O₄S₂
Molecular Weight: 268,3500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063095
CAS-Nummer: 462-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR