L-Cysteic acid 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-2-Amino-3-sulphopropanoic acid
OR480423-25G
APOSOR480423-25GEA
121
EUR
APOSOR480423-25G
L-Cysteic acid 97%
L-Cysteic acid
Formel:
C₃H₇NO₅S Molecular Weight: 169,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007524 CAS-Nummer: 498-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...