D-Propargylglycine 98%

Lieferant: Apollo Scientific

Synonyme: H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine

OR480427-5G
APOSOR480427-5GEA 165 EUR
APOSOR480427-5G
D-Propargylglycine 98%
D-Propargylglycine
Formel: C₅H₇NO₂
Molecular Weight: 113,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237407
CAS-Nummer: 23235-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR