2-Amino-2-(p-tolyl)propanoic acid

Lieferant: Apollo Scientific

OR480438-1G
APOSOR480438-1GEA 260 EUR
APOSOR480438-1G
2-Amino-2-(p-tolyl)propanoic acid
2-Amino-2-(p-tolyl)propanoic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662588
CAS-Nummer: 6269-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR