2-Amino-2-(1-naphthyl)acetic acid 96%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-2-(naphthalen-1-yl)acetic acid

OR480441-1G
APOSOR480441-1GEA 284 EUR
APOSOR480441-1G
2-Amino-2-(1-naphthyl)acetic acid 96%
2-Amino-2-(1-naphthyl)acetic acid
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201.221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662419
CAS-Nummer: 97611-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR