2-Amino-2-(2,3-dichlorophenyl)acetic acid

Lieferant: Apollo Scientific

OR480443-1G
APOSOR480443-1GEA 226 EUR
APOSOR480443-1G
2-Amino-2-(2,3-dichlorophenyl)acetic acid
2-Amino-2-(2,3-dichlorophenyl)acetic acid
Formel: C₈H₇Cl₂NO₂
Molecular Weight: 220,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662393
CAS-Nummer: 318270-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR