2-Amino-2-(4-isopropylphenyl)acetic acid

Lieferant: Apollo Scientific

OR480450-1G
APOSOR480450-1GEA 955 EUR
APOSOR480450-1G
2-Amino-2-(4-isopropylphenyl)acetic acid
2-Amino-2-(4-isopropylphenyl)acetic acid
Formel: C₁₁H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00665359
CAS-Nummer: 126746-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR