3-Chlor-L-tyrosin

Lieferant: Apollo Scientific

Synonyme: H-Tyr(3-Cl)-OH

OR480459-5G
APOSOR480459-5GEA 203 EUR
APOSOR480459-5G
3-Chlor-L-tyrosin
3-Chlor-L-tyrosin
Formel: C₉H₁₀ClNO₃
Molecular Weight: 215,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045865
CAS-Nummer: 7423-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR