2-Amino-2-(3-phenoxyphenyl)acetic acid 90%, tech

Lieferant: Apollo Scientific

OR480468-1G
APOSOR480468-1GEA 260 EUR
APOSOR480468-1G
2-Amino-2-(3-phenoxyphenyl)acetic acid 90%, tech
2-Amino-2-(3-phenoxyphenyl)acetic acid
Formel: C₁₄H₁₃NO₃
Molecular Weight: 243,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662402
CAS-Nummer: 299168-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR