2-Amino-2-(2-bromo-5-methoxy-phenyl)acetic acid 95%

Lieferant: Apollo Scientific

OR480469-1G
APOSOR480469-1GEA 496 EUR
APOSOR480469-1G
2-Amino-2-(2-bromo-5-methoxy-phenyl)acetic acid 95%
2-Amino-2-(2-bromo-5-methoxy-phenyl)acetic acid
Formel: C₉H₁₀BrNO₃
Molecular Weight: 260.085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04966784
CAS-Nummer: 1031705-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR