DL-Homoserin 95%

Lieferant: Apollo Scientific

OR480476-100G OR480476-25G
APOSOR480476-100GEA 0 EUR
APOSOR480476-100G APOSOR480476-25G
DL-Homoserin 95%
DL-Homoserin
Formel: C₄H₉NO₃
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002618
CAS-Nummer: 1927-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR