2-Bromo-4-methyl-5-nitroaniline 97%

Lieferant: Apollo Scientific

OR480479-5G
APOSOR480479-5GEA 282 EUR
APOSOR480479-5G
2-Bromo-4-methyl-5-nitroaniline 97%
2-Bromo-4-methyl-5-nitroaniline
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01239994
CAS-Nummer: 102169-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR