Isochinolin-5-carbonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyanoisoquinoline
OR48050-1G
APOSOR48050-1GEA
40.6
EUR
APOSOR48050-1G
Isochinolin-5-carbonitril
Isochinolin-5-carbonitril
Formel:
C₁₀H₆N₂ Molecular Weight: 154.1700 g/mol Schmelzpunkt: 135…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03109890 CAS-Nummer: 27655-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...