2-Amino-5-chlorobenzothiazole 97%

Lieferant: Apollo Scientific

Synonyme: 5-chloro-1,3-benzothiazol-2-amine

OR480503-5G
APOSOR480503-5GEA 27.7 EUR
APOSOR480503-5G
2-Amino-5-chlorobenzothiazole 97%
2-Amino-5-chlorobenzothiazole
Formel: C₇H₅ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00205355
CAS-Nummer: 20358-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR