2,4-Dichlorobenzothiazole 95+%

Lieferant: Apollo Scientific

OR480507-5G
APOSOR480507-5GEA 139 EUR
APOSOR480507-5G
2,4-Dichlorobenzothiazole 95+%
2,4-Dichlorobenzothiazole
Formel: C₇H₃Cl₂NS
Molecular Weight: 204,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127716
CAS-Nummer: 3622-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR