6-Amino-7-bromobenzothiazole 98%

Lieferant: Apollo Scientific

Synonyme: 7-bromo-1,3-benzothiazol-6-amine

OR480519-1G
APOSOR480519-1GEA 108 EUR
APOSOR480519-1G
6-Amino-7-bromobenzothiazole 98%
6-Amino-7-bromobenzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229,1000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426350
CAS-Nummer: 769-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR