2-Bromo-6-chlorobenzo[d]thiazole 97%
Lieferant: Apollo Scientific
OR480520-5G
APOSOR480520-5GEA
106
EUR
APOSOR480520-5G
2-Bromo-6-chlorobenzo[d]thiazole 97%
2-Bromo-6-chlorobenzo[d]thiazole
Formel:
C₇H₃BrClNS Lagertemperatur: Raumtemperatur |
MDL:
MFCD07783781 CAS-Nummer: 3507-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...