5-Methoxy-2,4(1H,3H)-pyrimidindion 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methoxy-2,4-pyrimidinediol

OR480522-25G
APOSOR480522-25GEA 71.2 EUR
APOSOR480522-25G
5-Methoxy-2,4(1H,3H)-pyrimidindion 95%
2,4-Dihydroxy-5-methoxypyrimidine
Formel: C₅H₆N₂O₃
Molecular Weight: 142,1100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00187903
CAS-Nummer: 6623-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR